Learn More
Invitrogen™ DDI2 Polyclonal Antibody

Description
DDI2 Polyclonal Antibody for Western Blot
Specifications
Specifications
| Antigen | DDI2 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.4 |
| Gene | DDI2 |
| Gene Accession No. | A2ADY9, Q5TDH0 |
| Gene Alias | 1110056G13Rik; 1700027M01Rik; 9130022E05Rik; AI604911; AU040698; DDI1, DNA-damage inducible 1, homolog 2; Ddi2; DNA damage inducible 1 homolog 2; DNA-damage inducible 1 homolog 2; DNA-damage inducible protein 2; DNA-damage inducible protein 2 (DDI2); protein DDI1 homolog 2; RP4-680D5.5 |
| Gene Symbols | DDI2 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.