Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DDIT3 Mouse anti-Human, Clone: 3H7, Abnova™

Catalog No. 89934038 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-934-038 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-934-038 Supplier Abnova Corporation Supplier No. MAB12865
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against full length recombinant human DDIT3.

Specifications

Antigen DDIT3
Applications Immunohistochemistry (PFA fixed), Western Blot
Classification Monoclonal
Clone 3H7
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against full length recombinant human DDIT3.
Dilution Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (10 ug/mL) Western Blot (1:2000) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.3 (50% glycerol, 1% BSA, 0.02% sodium azide).
Gene Alias CEBPZ/CHOP/CHOP10/GADD153/MGC4154
Gene Symbols DDIT3
Host Species Mouse
Immunogen Recombinant protein corresponding to full length human DDIT3.
Purification Method Protein A/G purification
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1649
Target Species Human
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.