Learn More
DDIT3 Mouse anti-Human, Clone: OTI4G2, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF802206

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
GADD153 is a small nuclear protein that is capable of dimerizing with transcription factors C/EBP alpha and beta. Once dimerized, this complex inhibits the normal binding and function of C/EBP to classical binding sites. Inversely, the C/EBP GADD153 dimer gains binding activity to other non classical C/EBP stress related targets. Under normal cellular conditions this protein is not expressed in detectable levels, but is highly unregulated during times of cellular/ER stress. Examples of GADD153 inducing stress include: treatment with tunicamycin, nutrient starvation and reducing agents that interfere with the calcium flux across the ER membrane.Specifications
DDIT3 | |
Monoclonal | |
Unconjugated | |
DDIT3 | |
C/EBPzeta, CEBPZ, CHOP, CHOP-10, CHOP10, GADD153 | |
Mouse | |
Affinity Chromatography | |
RUO | |
1649 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot | |
OTI4G2 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
P35638 | |
DDIT3 | |
Full length human recombit protein of human DDIT3 produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.