Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DDIT4, p Thr23, p Thr25 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP229521
Description
DDIT4 Polyclonal specifically detects DDIT4 in Human samples. It is validated for Western Blot.Specifications
DDIT4 (p Thr23, p Thr25) | |
Polyclonal | |
Western Blot 1:500 | |
Q9NX09 | |
DDIT4 | |
Synthetic phospho-peptide corresponding to amino acid residues surrounding Thr23/25 conjugated to KLH. Accession # Q9NX09 | |
Antigen Affinity-purified | |
RUO | |
Primary | |
Specific for the ∼34k REDD1 phosphorylated at Thr23/25. Immunolabeling is blocked by the phosphopeptide used as antigen but not by the corresponding dephosphopeptide. | |
Store at -20C. Avoid freeze-thaw cycles. |
Western Blot | |
Unconjugated | |
10mM HEPES (pH 7.5), 0.15M NaCl, 0.1 mg/ml BSA and 50% Glycerol with No Preservative | |
Dig2, DNA-damage-inducible transcript 4, FLJ20500, HIF-1 responsive RTP801 (RTP801), Protein regulated in development and DNA damage response 1, REDD-1DNA damage-inducible transcript 4 protein, REDD1HIF-1 responsive protein RTP801, RTP801RP11-442H21.1 | |
Rabbit | |
34 kDa | |
0.1 mL | |
Cancer, mTOR Pathway | |
54541 | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction