Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ DDIT4 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA520496
Description
A suggested positive control is 293 cell lysate. PA5-20496 can be used with blocking peptide PEP-0616.
DDIT4 gene ontology annotations related to this gene include cellular response to dexamethasone stimulus; intrinsic apoptotic signaling pathway in response to DNA damage by p53 class mediator; negative regulation of glycolytic process; negative regulation of TOR signaling; protein-containing complex disassembly; reactive oxygen species metabolic process; response to hypoxia.
Specifications
DDIT4 | |
Polyclonal | |
Unconjugated | |
DDIT4 | |
5830413E08Rik; AA415483; DDIT4; Dexamethasone-induced gene 2 protein; dig2; DNA damage inducible transcript 4; DNA damage-inducible transcript 4 protein; DNA-damage-inducible transcript 4; DNA-damage-inducible transcript 4 protein; HIF-1 responsive protein RTP801; HIF-1 responsive RTP801; protein regulated in development and DNA damage response 1; REDD1; REDD-1; RP11-442H21.1; RTP801 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
140942, 54541, 74747 | |
Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 0.02% sodium azide | |
Q8VHZ9, Q9D3F7, Q9NX09 | |
DDIT4 | |
A 14 amino acid peptide from near the amino terminus of human RTP801. | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction