Learn More
Invitrogen™ DDIT4 Polyclonal Antibody

Description
Positive Control: Hela and K562 cell lysates, rat epididymis tissue, human tonsil tissue, human colon cancer tissue, human breast tissue, mouse colon tissue, A549.
Specifications
Specifications
| Antigen | DDIT4 |
| Applications | Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol, 0.2% BSA and 0.05% sodium azide, pH 7.4 |
| Gene | DDIT4 |
| Gene Accession No. | Q9NX09 |
| Gene Alias | 5830413E08Rik; AA415483; DDIT4; Dexamethasone-induced gene 2 protein; dig2; DNA damage inducible transcript 4; DNA damage-inducible transcript 4 protein; DNA-damage-inducible transcript 4; DNA-damage-inducible transcript 4 protein; HIF-1 responsive protein RTP801; HIF-1 responsive RTP801; protein regulated in development and DNA damage response 1; REDD1; REDD-1; RP11-442H21.1; RTP801 |
| Gene Symbols | DDIT4 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.