Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DDIT4 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501986004
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-6004 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-6004 Supplier Bioss Supplier No. BS4240R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

DDIT4 gene ontology annotations related to this gene include cellular response to dexamethasone stimulus; intrinsic apoptotic signaling pathway in response to DNA damage by p53 class mediator; negative regulation of glycolytic process; negative regulation of TOR signaling; protein-containing complex disassembly; reactive oxygen species metabolic process; response to hypoxia.

Specifications

Antigen DDIT4
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene DDIT4
Gene Accession No. Q9NX09
Gene Alias 5830413E08Rik; AA415483; Ddit4; Dexamethasone-induced gene 2 protein; Dig2; DNA damage inducible transcript 4; DNA damage-inducible transcript 4 protein; DNA-damage-inducible transcript 4; DNA-damage-inducible transcript 4 protein; HIF-1 responsive protein RTP801; HIF-1 responsive RTP801; Protein regulated in development and DNA damage response 1; REDD1; REDD-1; RP11-442H21.1; RTP801
Gene Symbols DDIT4
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human DDIT4.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54541
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.