Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DDIT4L Antibody (DDIT-03), DyLight 650, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP262218C
Description
DDIT4L Monoclonal specifically detects DDIT4L in Human samples. It is validated for Western Blot, Flow Cytometry.Specifications
DDIT4L | |
Monoclonal | |
DyLight 650 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
DDIT4L | |
N-terminal recombinant fragment of human DDIT4L (amino acids 2-98) | |
0.1 mL | |
Primary | |
The antibody MEM-18 reacts with CD14, a 53-55 kDa GPI (glycosylphosphatidylinositol)-linked membrane glycoprotein expressed on monocytes, macrophages and weakly on granulocytes; also expressed by most tissue macrophages. In human, the epitope recognized by MEM-18 is located between amino acids 57-64. | |
Store at 4°C in the dark. |
Western Blot, Flow Cytometry | |
DDIT-03 | |
Western Blot, Flow Cytometry | |
DNA damage-inducible transcript 4-like protein, DNA-damage-inducible transcript 4-like, HIF-1 responsive protein RTP801-like, homolog of mouse SMHS1, Protein regulated in development and DNA damage response 2, REDD-2, REDD2Rtp801L, regulated in development and DNA damage response 2, RTP801L | |
Mouse | |
Protein A purified | |
Cancer, mTOR Pathway | |
115265 | |
Human | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction