Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DDIT4L Antibody (DDIT-03), mFluor Violet 610 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP262218MFV610
Description
DDIT4L Monoclonal antibody specifically detects DDIT4L in Human samples. It is validated for Flow Cytometry, Flow (Intracellular)Specifications
DDIT4L | |
Monoclonal | |
mFluor Violet 610 SE | |
50mM Sodium Borate | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry | |
DDIT-03 | |
Flow Cytometry, Flow (Intracellular) | |
DNA damage-inducible transcript 4-like protein, DNA-damage-inducible transcript 4-like, HIF-1 responsive protein RTP801-like, homolog of mouse SMHS1, Protein regulated in development and DNA damage response 2, REDD-2, REDD2Rtp801L, regulated in development and DNA damage response 2, RTP801L | |
N-terminal recombinant fragment of human DDIT4L (amino acids 2-98) | |
0.1 mL | |
Cancer, mTOR Pathway | |
115265 | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction