Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DDIT4L Monoclonal Antibody (DDIT-03)
GREENER_CHOICE

Catalog No. PIMA528629
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA528629 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA528629 Supplier Invitrogen™ Supplier No. MA528629
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This antibody recognizes DDIT4L / REDD2 protein, which belongs to intracellular stress-induced proteins involved in mediation of cell death.

DDIT4L inhibits cell growth by regulating the TOR signaling pathway upstream of the TSC1-TSC2 complex and downstream of AKT1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DDIT4L
Applications Flow Cytometry, Western Blot
Classification Monoclonal
Clone DDIT-03
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 15mM sodium azide; pH 7.4
Gene DDIT4L
Gene Accession No. Q96D03
Gene Alias 1700037B15Rik; 1700108M02Rik; Ddit4l; DNA damage inducible transcript 4 like; DNA damage-inducible transcript 4-like protein; DNA-damage-inducible transcript 4-like; DNA-damage-inducible transcript 4-like protein; DNA-damage-inducible transcript 4-like protein-like; HIF-1 responsive protein RTP801-like; homolog of mouse SMHS1; protein regulated in development and DNA damage response 2; REDD2; REDD-2; regulated in development and DNA damage response 2; RTP801L; skeletal muscle hindlimb suspension 1; Smhs1; Soleus muscle atrophied after hindlimb suspension protein 1
Gene Symbols DDIT4L
Host Species Mouse
Immunogen human DDIT4L (amino acids 2-98, N-terminal).
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 115265
Target Species Human
Content And Storage 4°C, do not freeze
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.