Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DDIT4L Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501987223
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-7223 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-7223 Supplier Bioss Supplier No. BS6376R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

DDIT4L inhibits cell growth by regulating the TOR signaling pathway upstream of the TSC1-TSC2 complex and downstream of AKT1.

Specifications

Antigen DDIT4L
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene DDIT4L
Gene Accession No. Q96D03
Gene Alias 1700037B15Rik; 1700108M02Rik; DDIT4L; DNA damage inducible transcript 4 like; DNA damage-inducible transcript 4-like protein; DNA-damage-inducible transcript 4-like; DNA-damage-inducible transcript 4-like protein; DNA-damage-inducible transcript 4-like protein-like; HIF-1 responsive protein RTP801-like; homolog of mouse SMHS1; Protein regulated in development and DNA damage response 2; REDD2; REDD-2; regulated in development and DNA damage response 2; RTP801L; skeletal muscle hindlimb suspension 1; Smhs1; Soleus muscle atrophied after hindlimb suspension protein 1
Gene Symbols DDIT4L
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human DDIT4L.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 115265
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.