Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ DDO Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA545190
Description
Peptide sequence: TQKQWFRETF NHLFAIANSA EAGDAGVHLV SGWQIFQSTP TEEVPFWADV Sequence homology: Cow: 86%; Dog: 93%; Horse: 93%; Human: 100%; Pig: 93%; Rabbit: 86%.
DDO is a peroxisomal flavoprotein that catalyzes the oxidative deamination of D-aspartate and N-methyl D-aspartate. Flavin adenine dinucleotide or 6-hydroxyflavin adenine dinucleotide can serve as the cofactor in this reaction.
Specifications
| DDO | |
| Polyclonal | |
| Unconjugated | |
| DDO | |
| 5330420D20Rik; 5730402C02Rik; AI467244; aspartic oxidase; DASOX; D-aspartate oxidase; D-aspartate oxidase, DDO; Ddo; DDO protein; DDO-1; DDO-2 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 8528 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q99489 | |
| DDO | |
| Synthetic peptide directed towards the N-terminal of human DDO (aa 63-112). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction