Learn More
Invitrogen™ DDOST Polyclonal Antibody

Description
Recommended positive controls: A431, H1299, HeLa, HepG2. Predicted reactivity: Mouse (96%), Rat (96%), Zebrafish (86%), Xenopus laevis (90%), Dog (97%), Pig (94%), Chicken (93%), Bovine (97%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Specifications
Specifications
| Antigen | DDOST |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.94 mg/mL |
| Conjugate | Unconjugated |
| Formulation | 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 |
| Gene | DDOST |
| Gene Accession No. | P39656 |
| Gene Alias | advanced glycation endproduct receptor 1; advanced glycation end-product receptor 1; AGER1; AGE-R1; CDG1R; Ddost; DDOST 48 kDa subunit; dolichyl-diphosphooligosaccharide protein glycosyltransferase; dolichyl-diphosphooligosaccharide-protein glycosyltransferase; dolichyl-diphosphooligosaccharide--protein glycosyltransferase 48 kDa subunit; dolichyl-diphosphooligosaccharide--protein glycosyltransferase non-catalytic subunit; dolichyl-diphosphooligosaccharide--protein glycosyltransferase subunit (non-catalytic); dolichyl-di-phosphooligosaccharide-protein glycotransferase; dolichyl-diphosphooligosaccharide-protein glycotransferase; KIAA0115; MGC2191; OK/SW-cl.45; OKSWcl45; oligosaccharyl transferase 48 kDa subunit; oligosaccharyltransferase 48 kDa subunit; oligosaccharyltransferase OST48; oligosaccharyltransferase subunit 48; OST; OST48; WBP1; wu:fa11d01; zgc:66068; zgc:66068 protein; zgc:77478 |
| Gene Symbols | DDOST |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.