Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DDOST Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA542898
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA542898 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA542898 Supplier Thermo Scientific Supplier No. PA542898
Add to Cart
Add to Cart

DDOST Polyclonal Antibody

DDOST Polyclonal Antibody for Western Blot

TRUSTED_SUSTAINABILITY

Specifications

Antigen DDOST
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene DDOST
Gene Accession No. P39656
Gene Alias advanced glycation endproduct receptor 1; advanced glycation end-product receptor 1; AGER1; AGE-R1; CDG1R; Ddost; DDOST 48 kDa subunit; dolichyl-diphosphooligosaccharide protein glycosyltransferase; dolichyl-diphosphooligosaccharide-protein glycosyltransferase; dolichyl-diphosphooligosaccharide--protein glycosyltransferase 48 kDa subunit; dolichyl-diphosphooligosaccharide--protein glycosyltransferase non-catalytic subunit; dolichyl-diphosphooligosaccharide--protein glycosyltransferase subunit (non-catalytic); dolichyl-di-phosphooligosaccharide-protein glycotransferase; dolichyl-diphosphooligosaccharide-protein glycotransferase; KIAA0115; MGC2191; OK/SW-cl.45; OKSWcl45; oligosaccharyl transferase 48 kDa subunit; oligosaccharyltransferase 48 kDa subunit; oligosaccharyltransferase OST48; oligosaccharyltransferase subunit 48; OST; OST48; WBP1; wu:fa11d01; zgc:66068; zgc:66068 protein; zgc:77478
Gene Symbols DDOST
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human DDOST (aa 101-150).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1650
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.