Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DDOST Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA597181
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA597181 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA597181 Supplier Invitrogen™ Supplier No. PA597181
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VCASGPRTLV LLDNLNVRET HSLFFRSLKD RGFELTFKTA DDPSLSLIKY GEFLYDNLII FSPSVEDFGG NINVETISAF IDGGGSVLVA ASSDIGDPLR ELGSECGIEF DEEKTAVIDH HNYDISDLGQ HTLIVADTEN LLKAPTIVGK SSLNPILFRG VGMVADPDNP LVLDILTGSS TSYSFFPDKP ITQYPHAVGK NTLLIAGLQA RNNARVIFSG SLDFFSDSFF NSAVQKAAPG SQRYSQTGNY ELAVALSRWV F; Positive Samples: U-87MG, HT-29, HepG2, A-549, Mouse kidney, Mouse liver, Mouse brain, Rat testis; Cellular Location: Endoplasmic reticulum membrane, Single-pass type I membrane protein.

This gene encodes a component of the oligosaccharyltransferase complex which catalyzes the transfer of high-mannose oligosaccharides to asparagine residues on nascent polypeptides in the lumen of the rough endoplasmic reticulum. The protein complex co-purifies with ribosomes. The product of this gene is also implicated in the processing of advanced glycation end products (AGEs), which form from non-enzymatic reactions between sugars and proteins or lipids and are associated with aging and hyperglycemia.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DDOST
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 2.88 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene DDOST
Gene Accession No. O54734, P39656, Q641Y0
Gene Alias advanced glycation endproduct receptor 1; advanced glycation end-product receptor 1; AGER1; AGE-R1; CDG1R; Ddost; DDOST 48 kDa subunit; dolichyl-diphosphooligosaccharide protein glycosyltransferase; dolichyl-diphosphooligosaccharide-protein glycosyltransferase; dolichyl-diphosphooligosaccharide--protein glycosyltransferase 48 kDa subunit; dolichyl-diphosphooligosaccharide--protein glycosyltransferase non-catalytic subunit; dolichyl-diphosphooligosaccharide--protein glycosyltransferase subunit (non-catalytic); dolichyl-di-phosphooligosaccharide-protein glycotransferase; dolichyl-diphosphooligosaccharide-protein glycotransferase; KIAA0115; MGC2191; OK/SW-cl.45; OKSWcl45; oligosaccharyl transferase 48 kDa subunit; oligosaccharyltransferase 48 kDa subunit; oligosaccharyltransferase OST48; oligosaccharyltransferase subunit 48; OST; OST48; WBP1; wu:fa11d01; zgc:66068; zgc:66068 protein; zgc:77478
Gene Symbols DDOST
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 40-300 of human DDOST (NP_0052072).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 13200, 1650, 313648
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.