Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DDR1 Antibody (203), Janelia Fluor™ 646 , Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP289618JF646
Description
DDR1 Monoclonal antibody specifically detects DDR1 in Human samples. It is validated for ELISASpecifications
DDR1 | |
Monoclonal | |
Janelia Fluor 646 | |
CD167 antigen-like family member A, CD167a antigen, DDR, discoidin domain receptor family, member 1, discoidin domain receptor tyrosine kinase 1, Discoidin receptor tyrosine kinase, EC 2.7.10, EC 2.7.10.1, EDDR1, ENTRK4, Epithelial discoidin domain receptor 1, HGK2, MCK10, MCK-10, NTRK4, Protein-tyrosine kinase 3A, PTK3A, PTK3A protein tyrosine kinase 3A, receptor, type 4, TRK E | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human DDR1 (NP_001945.3; Met 1-Thr 416). | |
0.1 mL | |
Cancer, Cellular Signaling | |
780 | |
Store at 4°C in the dark. | |
IgG |
ELISA | |
203 | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction