Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DDR1 Antibody (3E3), Alexa Fluor™ 350, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP281073AF350
Description
DDR1 Monoclonal specifically detects DDR1 in Human samples. It is validated for Western Blot, ELISA.Specifications
DDR1 | |
Monoclonal | |
Alexa Fluor 350 | |
50mM Sodium Borate | |
DDR1 | |
Recombinant human DDR1 extracellular domain. | |
0.1 mL | |
Primary | |
This antibody binds specifically to the extracellular domain of human DDR1. | |
Store at 4°C in the dark. | |
IgG κ |
Western Blot, ELISA | |
3000 | |
Western Blot, ELISA | |
CD167 antigen-like family member A, CD167a antigen, DDR, discoidin domain receptor family, member 1, discoidin domain receptor tyrosine kinase 1, Discoidin receptor tyrosine kinase, EC 2.7.10, EC 2.7.10.1, EDDR1, ENTRK4, Epithelial discoidin domain receptor 1, HGK2, MCK10, MCK-10, NTRK4, Protein-tyrosine kinase 3A, PTK3A, PTK3A protein tyrosine kinase 3A, receptor, type 4, TRK E | |
Rabbit | |
Protein A purified | |
RUO | |
780 | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction