Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DDR1 Antibody (4F2C12) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$584.50
Specifications
Antigen | DDR1 |
---|---|
Clone | 4F2C12 |
Concentration | 1 mg/mL |
Dilution | Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:1000, ELISA 1:100-1:2000 |
Applications | Western Blot, Flow Cytometry, ELISA |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP2-61692
![]() |
Novus Biologicals
NBP261692 |
0.1 mL |
Each of 1 for $584.50
|
|
|||||
Description
DDR1 Monoclonal specifically detects DDR1 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA.Specifications
DDR1 | |
1 mg/mL | |
Western Blot, Flow Cytometry, ELISA | |
Unconjugated | |
Mouse | |
780 | |
Purified recombinant fragment of human CD167 (AA: extra 21-176) expressed in E. Coli. | |
Primary | |
101 kDa |
4F2C12 | |
Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:1000, ELISA 1:100-1:2000 | |
Monoclonal | |
Purified | |
CD167 antigen-like family member A, CD167a antigen, DDR, discoidin domain receptor family, member 1, discoidin domain receptor tyrosine kinase 1, Discoidin receptor tyrosine kinase, EC 2.7.10, EC 2.7.10.1, EDDR1, ENTRK4, Epithelial discoidin domain receptor 1, HGK2, MCK10, MCK-10, NTRK4, Protein-tyrosine kinase 3A, PTK3A, PTK3A protein tyrosine kinase 3A, receptor, type 4, TRK E | |
DDR1 | |
IgG2b | |
Protein G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title