Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DDR1 Antibody (7F6), Novus Biologicals™

Human Monoclonal Antibody
$302.50 - $494.50
Specifications
Antigen | DDR1 |
---|---|
Clone | 7F6 |
Dilution | ELISA |
Applications | ELISA |
Classification | Monoclonal |
Description
DDR1 Monoclonal antibody specifically detects DDR1 in Human samples. It is validated for ELISASpecifications
DDR1 | |
ELISA | |
Monoclonal | |
Purified | |
RUO | |
Human | |
CD167 antigen-like family member A, CD167a antigen, DDR, discoidin domain receptor family, member 1, discoidin domain receptor tyrosine kinase 1, Discoidin receptor tyrosine kinase, EC 2.7.10, EC 2.7.10.1, EDDR1, ENTRK4, Epithelial discoidin domain receptor 1, HGK2, MCK10, MCK-10, NTRK4, Protein-tyrosine kinase 3A, PTK3A, PTK3A protein tyrosine kinase 3A, receptor, type 4, TRK E | |
Recombinant Human DDR1 protein [UniProt Q08345] | |
Primary | |
Store at -20 to -70C. Avoid freeze-thaw cycles. |
7F6 | |
ELISA | |
Unconjugated | |
Human | |
Cancer, Cellular Signaling | |
50% Glycerol, 0.01M PBS, PH 7.4 | |
780 | |
IgG1 | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title