Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ DDR1-IN-1 dihydrochloride
GREENER_CHOICESDP

Catalog No. 507710 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-771-0 10 mg
50-775-0R 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 50-771-0 Supplier Tocris Bioscience™ Supplier No. 5077/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Selective DDR1 inhibitor

DDR1-IN-1 dihydrochloride is a selective discoidin domain receptor 1 (DDR1) tyrosine kinase inhibitor (IC50 values are 105 and 413 nM for DDR1 and DDR2 respectively). Inhibits integrin-induced DDR1 autophosphorylation in an osteosarcoma cell line.

Specifications

CAS 1780303-76-5
Quantity 10 mg
Product Type DDR1-IN-1 dihydrochloride
Purity >98%
Molecular Weight (g/mol) 625.51
Inhibitors DDR1-IN-1 dihydrochloride

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.