Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DDR1, Rabbit anti-Human, Clone: 3000, Novus Biologicals™

Rabbit Monoclonal Antibody
$257.16 - $615.49
Specifications
| Antigen | DDR1 |
|---|---|
| Clone | 3000 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot, ELISA, Block/Neutralize |
| Applications | Western Blot, ELISA, Blocking Assay |
Description
DDR1 Monoclonal specifically detects DDR1 in Human samples. It is validated for Western Blot, ELISA, Block/Neutralize.Specifications
| DDR1 | |
| 1 mg/mL | |
| Western Blot, ELISA, Blocking Assay | |
| Unconjugated | |
| Rabbit | |
| Human | |
| CD167 antigen-like family member A, CD167a antigen, DDR, discoidin domain receptor family, member 1, discoidin domain receptor tyrosine kinase 1, Discoidin receptor tyrosine kinase, EC 2.7.10, EC 2.7.10.1, EDDR1, ENTRK4, Epithelial discoidin domain receptor 1, HGK2, MCK10, MCK-10, NTRK4, Protein-tyrosine kinase 3A, PTK3A, PTK3A protein tyrosine kinase 3A, receptor, type 4, TRK E | |
| DDR1 | |
| IgG κ | |
| Protein A purified | |
| This antibody binds specifically to the extracellular domain of human DDR1. |
| 3000 | |
| Western Blot, ELISA, Block/Neutralize | |
| Monoclonal | |
| Purified | |
| RUO | |
| PBS | |
| 780 | |
| Recombinant human DDR1 extracellular domain. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title