Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DDR2 Antibody (9A9), Novus Biologicals™

Rabbit Monoclonal Antibody
$302.50 - $494.50
Specifications
Antigen | DDR2 |
---|---|
Clone | 9A9 |
Dilution | Western Blot 1:500-1:2000, Flow Cytometry 1:50-1:200, ELISA |
Applications | Western Blot, Flow Cytometry, ELISA |
Classification | Monoclonal |
Description
DDR2 Monoclonal antibody specifically detects DDR2 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, ELISASpecifications
DDR2 | |
Western Blot 1:500-1:2000, Flow Cytometry 1:50-1:200, ELISA | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse | |
CD167 antigen-like family member B, CD167b antigen, Discoidin domain receptor 2, discoidin domain receptor family, member 2, discoidin domain receptor tyrosine kinase 2, discoidin domain-containing receptor 2, EC 2.7.10, EC 2.7.10.1, hydroxyaryl-protein kinase, migration-inducing gene 16 protein, neurotrophic tyrosine kinase receptor related 3, Neurotrophic tyrosine kinase, receptor-related 3, NTRKR3cell migration-inducing protein 20, Receptor protein-tyrosine kinase TKT, TKTMIG20a, TYRO10, Tyrosine-protein kinase TYRO10, tyrosylprotein kinase | |
A synthesized peptide derived from Human DDR2 [UniProt Q16832] | |
Primary | |
Store at -20 to -70C. Avoid freeze-thaw cycles. |
9A9 | |
Western Blot, Flow Cytometry, ELISA | |
Unconjugated | |
Rabbit | |
Cardiovascular Biology, CD Markers, Immunology, Neuroscience, Signal Transduction | |
PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
4921 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title