Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DDR2 Antibody (SR2306), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP321783100UL
Description
DDR2 Monoclonal antibody specifically detects DDR2 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
DDR2 | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot | |
SR2306 | |
Western Blot 1:500-1:2000 | |
CD167 antigen-like family member B, CD167b antigen, Discoidin domain receptor 2, discoidin domain receptor family, member 2, discoidin domain receptor tyrosine kinase 2, discoidin domain-containing receptor 2, EC 2.7.10, EC 2.7.10.1, hydroxyaryl-protein kinase, migration-inducing gene 16 protein, neurotrophic tyrosine kinase receptor related 3, Neurotrophic tyrosine kinase, receptor-related 3, NTRKR3cell migration-inducing protein 20, Receptor protein-tyrosine kinase TKT, TKTMIG20a, TYRO10, Tyrosine-protein kinase TYRO10, tyrosylprotein kinase | |
A synthesized peptide derived from human DDR2 (Uniprot #: Q16832) | |
100 μg | |
Cardiovascular Biology, CD Markers, Immunology, Neuroscience, Signal Transduction | |
4921 | |
Store at -20° C. Avoid freeze/thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction