Learn More
Promega DDR2 Kinase Enzyme System, 10microg
Catalog No. PRV4058 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
10 μg
Catalog No. Quantity
PRV4058 10 μg
1 options
Catalog No. PRV4058 Supplier Promega Supplier No. V4058
Recombinant human DDR2 (467-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. DDR2 is a member of a novel subclass of RTKs containing a distinct extracellular region encompassing a factor VIII-like domain.
  • Easily Screen and Profile DDR2 Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP−Glo™ Assay for bioluminescent detection of kinase activity
Specifications
Quantity 10 μg
Content And Storage <-65°C