Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DDR2 Polyclonal Antibody
DDR2 Polyclonal Antibody
Supplier: Thermo Scientific PA586976
Description
DDR2 Polyclonal Antibody for Western Blot

Specifications
DDR2 | |
Polyclonal | |
Unconjugated | |
Ddr2 | |
AW495251; CD_antigen: CD167b; CD167 antigen-like family member B; CD167b; CD167b antigen; cell migration-inducing protein 20; Ddr2; Discoidin domain receptor 2; discoidin domain receptor family, member 2; discoidin domain receptor tyrosine kinase 2; discoidin domain-containing receptor 2; Discoidin domain-containing receptor tyrosine kinase 2; hydroxyaryl-protein; hydroxyaryl-protein kinase; MIG20a; migration-inducing gene 16 protein; neurotrophic tyrosine kinase receptor related 3; neurotrophic tyrosine kinase, receptor-related 3; Ntrkr3; receptor protein-tyrosine kinase TKT; RP11-572K18.1; TKT; TYRO10; tyrosine-protein kinase TYRO 10; tyrosine-protein kinase TYRO10; tyrosylprotein kinase | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
18214, 4921, 685781 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ELISA, Western Blot | |
1 mg/mL | |
TBS with 1% BSA, 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
Q16832, Q62371 | |
Ddr2 | |
KLH conjugated synthetic peptide derived from human CD167b: 245-350. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction