Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DDR2 Recombinant Rabbit Monoclonal Antibody (102)
GREENER_CHOICE

Catalog No. PIMA531105
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA531105 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA531105 Supplier Invitrogen™ Supplier No. MA531105
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire. This antibody has specificity for Mouse DDR2/CD167b.

DDR2 is a member of a subclass of RTKs and contains a distinct extracellular region encompassing a factor VIII-like domain. DDR1 and DDR2 have been shown to be potently inhibited by Gleevec. DDR2 required for normal bone development. Regulates osteoblast differentiation and chondrocyte maturation via a signaling pathway that involves MAP kinases and leads to the activation of the transcription factor RUNX2. Regulates remodeling of the extracellular matrix by up-regulation of the collagenases MMP1, MMP2 and MMP13, and thereby facilitates cell migration and tumor cell invasion. Promotes fibroblast migration and proliferation, and thereby contributes to cutaneous wound healing.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DDR2
Applications ELISA
Classification Recombinant Monoclonal
Clone 102
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene Ddr2
Gene Accession No. Q62371
Gene Alias AW495251; CD_antigen: CD167b; CD167 antigen-like family member B; CD167b; CD167b antigen; cell migration-inducing protein 20; Ddr2; Discoidin domain receptor 2; discoidin domain receptor family, member 2; discoidin domain receptor tyrosine kinase 2; discoidin domain-containing receptor 2; Discoidin domain-containing receptor tyrosine kinase 2; hydroxyaryl-protein; hydroxyaryl-protein kinase; MIG20a; migration-inducing gene 16 protein; neurotrophic tyrosine kinase receptor related 3; neurotrophic tyrosine kinase, receptor-related 3; Ntrkr3; receptor protein-tyrosine kinase TKT; RP11-572K18.1; TKT; TYRO10; tyrosine-protein kinase TYRO 10; tyrosine-protein kinase TYRO10; tyrosylprotein kinase
Gene Symbols Ddr2
Host Species Rabbit
Immunogen Recombinant Mouse DDR2/CD167b Protein (Met1-Arg399).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18214
Target Species Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.