Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DDRGK1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA553049
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA553049 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA553049 Supplier Invitrogen™ Supplier No. PA553049
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ASAGQEPLHN EELAGAGRVA QPGPLEPEEP RAGGRPRRRR DLGSRLQAQR RAQRVAWAEA DENEEEAVIL Highest antigen sequence identity to the following orthologs: Mouse - 68%, Rat - 70%.

Protein which interacts with the E3 UFM1-protein ligase UFL1 and one of its substrates TRIP4 and is required for TRIP4 ufmylation. Through TRIP4 ufmylation may regulate nuclear receptors-mediated transcription. May play a role in NF-kappa-B-mediated transcription through regulation of the phosphorylation and the degradation of NFKBIA, the inhibitor of NF-kappa-B. May also play a role in the cellular response to endoplasmic reticulum stress.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DDRGK1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene DDRGK1
Gene Accession No. Q96HY6
Gene Alias 1110001I20Rik; 2600009E05Rik; AI326138; C13H20ORF116; C20orf116; Dashurin; DDRGK domain containing 1; DDRGK domain-containing protein 1; Ddrgk1; dJ1187M17.3; RGD1309979; UFBP1; UFM1-binding and PCI domain-containing protein 1; UFM1-binding protein 1 containing a PCI domain
Gene Symbols DDRGK1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human DDRGK1. Recombinant protein control fragment (Product #RP-90580).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 65992
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.