Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DDRGK1 Rabbit anti-Human, Polyclonal, Bioss

Rabbit Polyclonal Antibody
Supplier: Bioss BS8252R

Description
Protein which interacts with the E3 UFM1-protein ligase UFL1 and one of its substrates TRIP4 and is required for TRIP4 ufmylation. Through TRIP4 ufmylation may regulate nuclear receptors-mediated transcription. May play a role in NF-kappa-B-mediated transcription through regulation of the phosphorylation and the degradation of NFKBIA, the inhibitor of NF-kappa-B. May also play a role in the cellular response to endoplasmic reticulum stress.Specifications
DDRGK1 | |
Polyclonal | |
Unconjugated | |
DDRGK1 | |
1110001I20Rik; 2600009E05Rik; AI326138; BOS_13369; C13H20ORF116; C20orf116; Dashurin; DDRGK domain containing 1; DDRGK domain containing 1-like; DDRGK domain-containing protein 1; DDRGK1; dJ1187M17.3; LOC100344812; Ufbp1; UFM1-binding and PCI domain-containing protein 1; UFM1-binding protein 1 containing a PCI domain | |
Rabbit | |
Protein A | |
RUO | |
65992 | |
-20°C | |
Liquid |
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
1 μg/mL | |
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH | |
Q96HY6 | |
DDRGK1 | |
KLH conjugated synthetic peptide derived from human DDRGK1. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction