Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DDT Antibody (OTI1H3), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP245677
Description
Ensure accurate, reproducible results in Western Blot, Immunohistochemistry
DDT Monoclonal specifically detects DDT in Human, Monkey samples. It is validated for Western Blot, Immunohistochemistry.Specifications
DDT | |
Monoclonal | |
1 mg/mL | |
Western Blot 1:200 - 8000, Immunohistochemistry 1:150, Immunohistochemistry-Paraffin 1:150 | |
D-dopachrome decarboxylase, D-dopachrome tautomeraseDDCT, EC 4.1.1.84, Phenylpyruvate tautomerase II | |
Mouse | |
12.5 kDa | |
0.1 mL | |
Stem Cell Markers | |
1652 | |
Store at -20C. Avoid freeze-thaw cycles. | |
IgG1 |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
OTI1H3 | |
Unconjugated | |
PBS (pH 7.3), 1.0% BSA and 50% Glycerol with 0.02% Sodium Azide | |
DDT | |
Full length human recombinant protein of human DDT(NP_001346) produced in HEK293T cell. | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Monkey | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction