Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DDT Monoclonal Antibody (OTI1A1)
GREENER_CHOICE

Catalog No. PIMA526175
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA526175 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA526175 Supplier Invitrogen™ Supplier No. MA526175
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

DDT Monoclonal Antibody for Western Blot, IHC (P)

Tautomerization of D-dopachrome with decarboxylation to give 5,6-dihydroxyindole (DHI).
TRUSTED_SUSTAINABILITY

Specifications

Antigen DDT
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI1A1
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene DDT
Gene Accession No. P30046
Gene Alias C78655; DDCT; D-dopachrome decarboxylase; D-dopachrome tautomerase; Ddt; dopachrome isomerase; hCG_41098; phenylpyruvate tautomerase II
Gene Symbols DDT
Host Species Mouse
Immunogen Full length human recombinant protein of DDT produced in HEK293T cell.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1652
Target Species Human, Monkey
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.