Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ DDT Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA521386
Description
Recommended positive controls: 293T, HeLa, Raji, mouse liver, rat liver. Predicted reactivity: Pig (80%), Rhesus Monkey (96%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Tautomerization of D-dopachrome with decarboxylation to give 5,6-dihydroxyindole (DHI).
Specifications
DDT | |
Polyclonal | |
Unconjugated | |
DDT | |
C78655; DDCT; D-dopachrome decarboxylase; D-dopachrome tautomerase; Ddt; dopachrome isomerase; hCG_41098; phenylpyruvate tautomerase II | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
13202, 1652, 29318 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
O35215, P30046, P80254 | |
DDT | |
Recombinant protein encompassing a sequence within the center region of human DDT. The exact sequence is proprietary. | |
100 μL | |
Primary | |
Human, Mouse, Rat, Canine | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction