Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DDT Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA597160
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA597160 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA597160 Supplier Invitrogen™ Supplier No. PA597160
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MPFLELDTNL PANRVPAGLE KRLCAAAASI LGKPADRVNV TVRPGLAMAL SGSTEPCAQL SISSIGVVGT AEDNRSHSAH FFEFLTKELA LGQDRILIRF FPLESWQIGK IGTVMTFL; Positive Samples: 293T, A-549, Mouse liver, Rat liver; Cellular Location: Cytoplasm.

Tautomerization of D-dopachrome with decarboxylation to give 5,6-dihydroxyindole (DHI).
TRUSTED_SUSTAINABILITY

Specifications

Antigen DDT
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 3.36 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene DDT
Gene Accession No. O35215, P30046, P80254
Gene Alias C78655; DDCT; D-dopachrome decarboxylase; D-dopachrome tautomerase; Ddt; dopachrome isomerase; hCG_41098; phenylpyruvate tautomerase II
Gene Symbols DDT
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-118 of human DDT (NP_0013461).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 13202, 1652, 29318
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.