Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DDX1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $646.00
Specifications
| Antigen | DDX1 |
|---|---|
| Dilution | Western Blot 0.4 ug/ml, Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:50 - 1:200 |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
DDX1 Polyclonal specifically detects DDX1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| DDX1 | |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| Human, Mouse, Rat | |
| ATP-dependent RNA helicase DDX1, DBP-RBDEAD box polypeptide 1, DEAD (Asp-Glu-Ala-Asp) box polypeptide 1, DEAD box protein 1, DEAD box protein retinoblastoma, DEAD box-1, DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 1, EC 3.6.1, EC 3.6.1.10, EC 3.6.4.13, UKVH5d | |
| DDX1 | |
| IgG | |
| Affinity Purified | |
| Specificity of human DDX1 antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Western Blot 0.4 ug/ml, Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| Polyclonal | |
| Rabbit | |
| Cancer, Neuroscience, Transcription Factors and Regulators, Vision | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| 1653 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:NQALFPACVLKNAELKFNFGEEEFKFPPKDGFVALSKAPDGYIVKSQHSGNAQVTQTKFLPNAPKALIVEPSRELAEQTLNNIKQFKKY | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title