Learn More
Invitrogen™ DDX1 Polyclonal Antibody

Description
Recommended positive controls: A431, H1299, HeLa, HepG2. Predicted reactivity: Mouse (97%), Rat (98%), Zebrafish (89%), Xenopus laevis (88%), Chicken (93%), Rhesus Monkey (98%), Bovine (98%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Specifications
Specifications
| Antigen | DDX1 |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry, Western Blot |
| Classification | Polyclonal |
| Concentration | 0.76 mg/mL |
| Conjugate | Unconjugated |
| Formulation | 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 |
| Gene | DDX1 |
| Gene Accession No. | Q641Y8, Q91VR5, Q92499 |
| Gene Alias | AA409185; ATP-dependent RNA helicase DDX1; DBP-RB; Ddx1; DEAD (Asp-Glu-Ala-Asp) box helicase 1; DEAD (Asp-Glu-Ala-Asp) box polypeptide 1; DEAD box polypeptide 1; DEAD box protein 1; DEAD box protein retinoblastoma; DEAD box-1; DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 1; DEAD/H-box helicase 1; UKVH5d |
| Gene Symbols | DDX1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.