Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DDX10 Antibody, DyLight 650, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB1001576C
Description
DDX10 Polyclonal antibody specifically detects DDX10 in Human samples. It is validated for Western BlotSpecifications
DDX10 | |
Polyclonal | |
Western Blot | |
DDX10-NUP98 fusion protein type 2, DEAD (Asp-Glu-Ala-Asp) box polypeptide 10, DEAD box protein 10, DEAD box-10, DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 10 (RNA helicase), EC 3.6.1, EC 3.6.4.13, HRH-J8, probable ATP-dependent RNA helicase DDX10 | |
The immunogen recognized by this antibody maps to a region between residues 500 and 550 of human DEAD (Asp-Glu-Ala-Asp) Box Polypeptide 10 using the numbering given in entry NP_004389.2 (GeneID 1662). | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot | |
DyLight 650 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
1662 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction