Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DDX11 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501987954
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-7954 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-7954 Supplier Bioss Supplier No. BS7719R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

DEAD box proteins, characterized by the conserved motifAsp-Glu-Ala-Asp (DEAD), are putative RNA helicases. They areimplicated in a number of cellular processes involving alterationof RNA secondary structure such as translation initiation, nuclearand mitochondrial splicing, and ribosome and spliceosome assembly.Based on their distribution patterns, some members of this familyare believed to be involved in embryogenesis, spermatogenesis, andcellular growth and division. DDX11 encodes a DEAD box protein,which is an enzyme that possesses both ATPase and DNA helicaseactivities. DDX11 is a homolog of the yeast CHL1 gene, and mayfunction to maintain chromosome transmission fidelity and genomestability.

Specifications

Antigen DDX11
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene DDX11
Gene Accession No. Q96FC9
Gene Alias 4732462I11Rik; ATP-dependent DNA helicase DDX11; CHL1; CHL1-like helicase homolog; CHL1-related helicase gene-1; CHL1-related protein 1; CHLR1; DDX11; DEAD/H (Asp-Glu-Ala-Asp/His) box helicase 11; DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 11 (CHL1-like helicase homolog, S. cerevisiae); DEAD/H box protein 11; DEAD/H-box helicase 11; DEAD/H-box protein 11; essa15a; hCHLR1; Keratinocyte growth factor-regulated gene 2 protein; KRG2; KRG-2; probable ATP-dependent DNA helicase DDX11; probable ATP-dependent RNA helicase DDX11; WABS
Gene Symbols DDX11
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human DDX11.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1663
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.