Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DDX18 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA523463
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA523463 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA523463 Supplier Invitrogen™ Supplier No. PA523463
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

DDX18 Polyclonal Antibody for Western Blot, IHC (P)

ATP-dependent RNA helicase DDX18 belongs to theDEAD box helicase family, which contains one helicase ATP-binding domain and one helicase C-terminal domain. DDX18 is induced by Myc and Max and is involved in splicing and nuclear export. It is reported to be differentially expressed during viral latency and at early times after induction into active replication.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DDX18
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% BSA and 0.05% sodium azide
Gene DDX18
Gene Accession No. Q9NVP1
Gene Alias ATP-dependent RNA helicase DDX18; cPERP-D; DDX18; DEAD (Asp-Glu-Ala-Asp) box polypeptide 18; DEAD box protein 18; DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 18 (Myc-regulated); DEAD-box helicase 18; MrDb; Myc-regulated DEAD box protein
Gene Symbols DDX18
Host Species Rabbit
Immunogen A portion of amino acids 20-80 of human DDX18.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8886
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.