Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DDX19A Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.00 - $487.50
Specifications
| Antigen | DDX19A |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Purified |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP15720720
![]() |
Novus Biologicals
NBP15720720UL |
20 μL |
Each for $206.00
|
|
|||||
NBP157207
![]() |
Novus Biologicals
NBP157207 |
100 μL |
Each for $487.50
|
|
|||||
Description
DDX19A Polyclonal specifically detects DDX19A in Human samples. It is validated for Western Blot.Specifications
| DDX19A | |
| Polyclonal | |
| Purified | |
| RUO | |
| ATP-dependent RNA helicase DDX19A, DDX19-DDX19L, DDX19L, DDX19-like protein, DEAD (Asp-Glu-Ala-As) box polypeptide 19A, DEAD (Asp-Glu-Ala-As) box polypeptide 19-like, DEAD box protein 19A, DKFZp686C21137, EC 3.6.1, EC 3.6.4.13, FLJ11126, RNA helicase | |
| DDX19A | |
| IgG | |
| Protein A purified |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Q9NUU7 | |
| 55308 | |
| Synthetic peptides corresponding to DDX19A (DEAD (Asp-Glu-Ala-As) box polypeptide 19A) The peptide sequence was selected from the N terminal of DDX19A. Peptide sequence IKEEKVKADTNGIIKTSTTAEKTDEEEKEDRAAQSLLNKLIRSNLVDNTN. | |
| Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title