Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DDX19A Monoclonal Antibody (OTI3C12)
GREENER_CHOICE

Catalog No. PIMA527239
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA527239 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA527239 Supplier Invitrogen™ Supplier No. MA527239
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

DDX19A Monoclonal Antibody for Western Blot, IHC (P)

ATP-dependent RNA helicase involved in mRNA export from the nucleus.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DDX19A
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI3C12
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene DDX19A
Gene Accession No. Q9NUU7
Gene Alias ATP-dependent RNA helicase DDX19A; DBP5; Ddx19; DDX19A; DDX19-DDX19L; DDX19L; DDX19-like protein; DEAD (Asp-Glu-Ala-As) box polypeptide 19A; DEAD (Asp-Glu-Ala-Asp) box polypeptide 19; DEAD (Asp-Glu-Ala-Asp) box polypeptide 19a; DEAD box protein 19A; DEAD box RNA helicase DEAD5; DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 19; DEAD-box helicase 19A; Eif4a-rs1; eukaryotic translation initiation factor 4A related sequence 1; eukaryotic translation initiation factor 4A-related sequence 1; mDEAD5; RNA helicase; ZD10B; zinc responsive protein ZD10B
Gene Symbols DDX19A
Host Species Mouse
Immunogen Full length human recombinant protein of DDX19A produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55308
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.