Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DDX19A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA541284
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA541284 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA541284 Supplier Invitrogen™ Supplier No. PA541284
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: IKEEKVKADT NGIIKTSTTA EKTDEEEKED RAAQSLLNKL IRSNLVDNTN Sequence homology: Dog: 85%; Horse: 86%; Human: 100%; Mouse: 93%; Rat: 93%.

ATP-dependent RNA helicase involved in mRNA export from the nucleus.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DDX19A
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene DDX19A
Gene Accession No. Q9NUU7
Gene Alias ATP-dependent RNA helicase DDX19A; DBP5; Ddx19; DDX19A; DDX19-DDX19L; DDX19L; DDX19-like protein; DEAD (Asp-Glu-Ala-As) box polypeptide 19A; DEAD (Asp-Glu-Ala-Asp) box polypeptide 19; DEAD (Asp-Glu-Ala-Asp) box polypeptide 19a; DEAD box protein 19A; DEAD box RNA helicase DEAD5; DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 19; DEAD-box helicase 19A; Eif4a-rs1; eukaryotic translation initiation factor 4A related sequence 1; eukaryotic translation initiation factor 4A-related sequence 1; mDEAD5; RNA helicase; ZD10B; zinc responsive protein ZD10B
Gene Symbols DDX19A
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human DDX19A (aa 25-74).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55308
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.