Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ DDX19A Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA531264
Description
Recommended positive controls: NT2D1, IMR32, U87-MG. Predicted reactivity: Mouse (100%), Rat (100%), Bovine (98%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
ATP-dependent RNA helicase involved in mRNA export from the nucleus.
Specifications
| DDX19A | |
| Polyclonal | |
| Unconjugated | |
| DDX19A | |
| ATP-dependent RNA helicase DDX19A; DBP5; Ddx19; DDX19A; DDX19-DDX19L; DDX19L; DDX19-like protein; DEAD (Asp-Glu-Ala-As) box polypeptide 19A; DEAD (Asp-Glu-Ala-Asp) box polypeptide 19; DEAD (Asp-Glu-Ala-Asp) box polypeptide 19a; DEAD box protein 19A; DEAD box RNA helicase DEAD5; DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 19; DEAD-box helicase 19A; Eif4a-rs1; eukaryotic translation initiation factor 4A related sequence 1; eukaryotic translation initiation factor 4A-related sequence 1; mDEAD5; RNA helicase; ZD10B; zinc responsive protein ZD10B | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 55308 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
| Q9NUU7 | |
| DDX19A | |
| Recombinant protein encompassing a sequence within the C-terminus region of human DDX19A. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction