Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DDX19B Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 4 publications
Supplier: Novus Biologicals NB100760
Description
DDX19B Polyclonal specifically detects DDX19B in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin.Specifications
DDX19B | |
Polyclonal | |
Unconjugated | |
TBS and 0.1% BSA with 0.09% Sodium Azide | |
ATP-dependent RNA helicase DDX19, DBP5ATP-dependent RNA helicase DDX19B, DDX19DEAD-box protein 5, DEAD (Asp-Glu-Ala-As) box polypeptide 19, DEAD (Asp-Glu-Ala-As) box polypeptide 19B, DEAD box protein 19B, DEAD box RNA helicase DEAD5, DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 19 (Dbp5, yeast, homolog), DEAD-box RNA helicase DEAD5, EC 3.6.1, EC 3.6.4.13, RNAh, yeast Dbp5 homolog | |
Rabbit | |
Affinity Purified | |
RUO | |
11269 | |
Human, Mouse | |
IgG |
Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
0.2 mg/mL | |
Western Blot 1:1000-1:10000, Immunohistochemistry 1:100-1:500, Immunoprecipitation 2- 5 ug/mg lysate, Immunohistochemistry-Paraffin 1:100-1:500 | |
NP_009173.1 | |
DDX19B | |
The immunogen recognized by this antibody maps to a region between residue 450 and the C-terminus (residue 479) of human DEAD (Asp-Glu-Ala-Asp) box polypeptide 19B using the numbering given in entry NP_009173.1 (GeneID 11269). | |
0.1 mL | |
Primary | |
Based on 100% sequence identity, this antibody is predicted to react with X. laevis, X. tropicalis, Orangutan, Root node of taxonomy, Treponema denticola, Zymomonas mobilis, Vibrio parahaemolyticus, Prochlorococcus marinus and Baker's yeast. | |
Store at 4C. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction