Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ DDX19B Polyclonal Antibody

Goat Polyclonal Antibody
Supplier: Invitrogen™ PA518755
Description
This antibody is predicted to react with canine, mouse and rat based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 4,000.
Spectrins are a family of filamentous cytoskeletal proteins that function as essential scaffold proteins that stabilize the plasma membrane and organize intracellular organelles. Spectrins are composed of alpha and beta dimers that associate to form tetramers linked in a head-to-head arrangement. This gene encodes an alpha spectrin that is specifically expressed in nonerythrocytic cells. The encoded protein has been implicated in other cellular functions including DNA repair and cell cycle regulation. Mutations in this gene are the cause of early infantile epileptic encephalopathy-5. Alternate splicing results in multiple transcript variants.
Specifications
| DDX19B | |
| Polyclonal | |
| Unconjugated | |
| DDX19B | |
| 2810457M08Rik; 4921519L13Rik; ATP-dependent RNA helicase DDX19; ATP-dependent RNA helicase DDX19B; AW260119; DBP5; DDX19; Ddx19b; DEAD (Asp-Glu-Ala-As) box polypeptide 19B; DEAD (Asp-Glu-Ala-Asp) box polypeptide 19b; DEAD box protein 19B; DEAD box RNA helicase DEAD5; DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 19 (Dbp5, yeast, homolog); DEAD-box helicase 19B; DEAD-box protein 5; DEAD-box RNA helicase DEAD5; RNAh; TDBP; Unknown (protein for MGC:134354); yeast Dbp5 homolog; Zd10a; zinc responsive protein Zd10A | |
| Goat | |
| Ammonium Sulfate Precipitation | |
| RUO | |
| 11269 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| Q9UMR2 | |
| DDX19B | |
| Synthetic peptide sequence (DLPVDKDGNPDNETY) corresponding to the internal amino acids of DDX19B (aa 409-424). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction