Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DDX21 Antibody, PE/Cy5.5, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB1001718PECY55
Description
DDX21 Polyclonal specifically detects DDX21 in Human, Mouse, Zebrafish samples. It is validated for Flow Cytometry.Specifications
DDX21 | |
Polyclonal | |
Flow Cytometry | |
DEAD (Asp-Glu-Ala-Asp) box polypeptide 21, DEAD box protein 21, DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 21, DKFZp686F21172, EC 3.6.1, EC 3.6.4.13, Gu protein, GUA, gu-alpha, GURDB, nucleolar RNA helicase 2, Nucleolar RNA helicase Gu, Nucleolar RNA helicase II, RH II/Gu, RH-II/GU, RH-II/GuA, RNA helicase II/Gu alpha | |
Rabbit | |
Affinity Purified | |
Primary | |
Human, Mouse, Zebrafish | |
IgG |
KnockDown | |
PE-Cyanine5.5 | |
PBS with 0.05% Sodium Azide | |
DDX21 | |
The immunogen recognized by this antibody maps to a region between residue 725 and the C-terminus (residue 783) of human DEAD/H (Asp-Glu-Ala-Asp/His) Box Polypeptide 21 using the numbering given in entry NP_004719.2 (GeneID 9188). | |
0.1 mL | |
9188.0 | |
Store at 4°C in the dark. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction