Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DDX21 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA541267
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA541267 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA541267 Supplier Thermo Scientific Supplier No. PA541267
Add to Cart
Add to Cart

DDX21 Polyclonal Antibody

DDX21 Polyclonal Antibody for Western Blot

TRUSTED_SUSTAINABILITY

Specifications

Antigen DDX21
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene DDX21
Gene Accession No. Q9NR30
Gene Alias AI255159; AL022742; D10Ertd645e; D10Wsu42e; DDX21; Ddx21a; DEAD (Asp-Glu-Ala-Asp) box helicase 21; DEAD (Asp-Glu-Ala-Asp) box polypeptide 21; DEAD (Asp-Glu-Ala-Asp) box polypeptide 21a; DEAD box protein 21; DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 21; DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 21 (RNA helicase II/Gu); DEAD-box helicase 21; DExD-box helicase 21; Gu protein; GUA; gu-alpha; GURDB; Nucleolar RNA helicase 2; nucleolar RNA helicase Gu; Nucleolar RNA helicase II; RH II/Gu; RH-II/GU; RH-II/GuA; RH-II/Gualpha; RNA helicase II/Gu alpha
Gene Symbols DDX21
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C-terminal of human DDX21 (aa 734-783).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9188
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.