Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DDX23 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP216130
Description
DDX23 Polyclonal specifically detects DDX23 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
DDX23 | |
Polyclonal | |
Unconjugated | |
PBS (pH 7.0), 1.0% BSA and 20% Glycerol with 0.01% Thimerosal | |
100 kDa U5 snRNP-specific protein, DEAD (Asp-Glu-Ala-Asp) box polypeptide 23, DEAD box protein 23, EC 3.6.1, EC 3.6.4.13, MGC8416, probable ATP-dependent RNA helicase DDX23, prp28, PRP28 homolog, PRP28 homolog, yeast, PRP28p homolog, PRPF28, U5 snRNP 100 kD protein, U5-100K, U5-100kD | |
Rabbit | |
96 kDa | |
0.1 mL | |
Cell Cycle and Replication | |
9416 | |
Human, Mouse | |
IgG |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
1mg/mL | |
Western Blot 1:500-1:3000, Immunohistochemistry 1:100-1:1000, Immunocytochemistry/Immunofluorescence 1:100-1:1000, Immunohistochemistry-Paraffin 1:100-1:1000 | |
Q9BUQ8 | |
DDX23 | |
Recombinant protein encompassing a sequence within the center region of human DDX23. The exact sequence is proprietary. | |
Affinity Purified | |
RUO | |
Primary | |
Expected to cross react based on sequence identity: Zebrafish (86%). | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction