Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DDX23 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA530312
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA530312 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA530312 Supplier Invitrogen™ Supplier No. PA530312
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: Jurkat, NIH-3T3, JC. Predicted reactivity: Mouse (100%), Rat (100%), Zebrafish (91%), Rhesus Monkey (100%), Bovine (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

PC4 (activated RNA polymerase II transcription cofactor 4) is a transcriptional coactivator, possessing the ability to suppress promoter-driven as well as nonspecific transcription via its DNA binding activity. The repressive activity of PC4 on promoter-driven transcription is alleviated by transcription factor TFIIH. TFIIH protects promoters from PC4-mediated repression by relieving the topological constraint imposed by PC4 through the ERCC3 helicase activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DDX23
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
Gene Ddx23
Gene Accession No. Q9BUQ8
Gene Alias 100 kDa U5 snRNP-specific protein; 3110082M05Rik; 4921506D17Rik; DDX23; DEAD (Asp-Glu-Ala-Asp) box polypeptide 23; DEAD box protein 23; DEAD-box helicase 23; probable ATP-dependent RNA helicase DDX23; prp28; PRP28 homolog; PRP28 homolog, yeast; PRP28p homolog; PRPF28; SNRNP100; U5 snRNP 100 kD protein; U5-100K; U5-100KD
Gene Symbols Ddx23
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human DDX23. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 74351, 9416
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.