Learn More
DDX23 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
$151.31 - $568.26
Specifications
Antigen | DDX23 |
---|---|
Applications | Immunohistochemistry |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Liquid |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-157-2220
![]() |
Bethyl Laboratories, Inc
IHC00511T |
10 μL |
Each for $151.31
|
|
|||||
50-157-2219
![]() |
Bethyl Laboratories, Inc
IHC00511 |
100 μL |
Each for $568.26
|
|
|||||
Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Mouse, Rat, Bovine, Dog, Horse, Rabbit, Guinea pig_10141, Pig, Panda, Orangutan, Gorilla a
Involved in pre-mRNA splicing and its phosphorylated form (by SRPK2) is required for spliceosomal B complex formation.Specifications
DDX23 | |
Polyclonal | |
Liquid | |
RUO | |
TBS with 0.1% BSA and 0.09% sodium azide | |
PRPF28, U5-100K, U5-100KD, prp28, 100 kDa U5 snRNP-specific protein, DEAD box protein 23, PRP28 homolog, yeast, PRP28p homolog, U5 snRNP 100 kD protein, probable ATP-dependent RNA helicase DDX23 | |
DDX23 | |
IgG | |
Antigen affinity chromatography | |
Antibody |
Immunohistochemistry | |
Unconjugated | |
Rabbit | |
Human | |
Q9BUQ8 | |
9416 | |
Between 1 and 50 | |
Primary | |
4° C | |
DDX23 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.