Learn More
DDX23 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
$167.95
Specifications
| Antigen | DDX23 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Liquid |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-155-8238
|
Bethyl Laboratories, Inc
A300695AT |
20 μL |
Each of 1 for $167.95
|
|
|||||
Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Orangutan
Involved in pre-mRNA splicing and its phosphorylated form (by SRPK2) is required for spliceosomal B complex formation.Specifications
| DDX23 | |
| Polyclonal | |
| Liquid | |
| RUO | |
| TBS with 0.1% BSA and 0.09% sodium azide | |
| PRPF28, U5-100K, U5-100KD, prp28, 100 kDa U5 snRNP-specific protein, DEAD box protein 23, PRP28 homolog, yeast, PRP28p homolog, U5 snRNP 100 kD protein, probable ATP-dependent RNA helicase DDX23 | |
| Between 775 and C-term | |
| Primary | |
| 4° C | |
| DDX23 |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse | |
| Q9BUQ8 | |
| 74351, 9416 | |
| IgG | |
| Antigen affinity chromatography | |
| Antibody |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.