Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ DDX23 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA583446
Description
Immunogen sequence: REEKDKSKEL HAIKERYLGG IKKRRRTRHL NDRKFVFEWD ASEDTSIDYN PLYKERHQVQ LLGRGFIAGI DLKQQKREQS RFYGDLM.
PC4 (activated RNA polymerase II transcription cofactor 4) is a transcriptional coactivator, possessing the ability to suppress promoter-driven as well as nonspecific transcription via its DNA binding activity. The repressive activity of PC4 on promoter-driven transcription is alleviated by transcription factor TFIIH. TFIIH protects promoters from PC4-mediated repression by relieving the topological constraint imposed by PC4 through the ERCC3 helicase activity.
Specifications
DDX23 | |
Polyclonal | |
Unconjugated | |
Ddx23 | |
100 kDa U5 snRNP-specific protein; 3110082M05Rik; 4921506D17Rik; DDX23; DEAD (Asp-Glu-Ala-Asp) box polypeptide 23; DEAD box protein 23; DEAD-box helicase 23; probable ATP-dependent RNA helicase DDX23; prp28; PRP28 homolog; PRP28 homolog, yeast; PRP28p homolog; PRPF28; SNRNP100; U5 snRNP 100 kD protein; U5-100K; U5-100KD | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
9416 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot, Immunocytochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9BUQ8 | |
Ddx23 | |
Recombinant protein corresponding to Human DDX23. Recombinant protein control fragment (Product #RP-104698). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction